Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

869008-M

Sigma-Aldrich

Growth Hormone, Human Pituitary, Iodination Grade

Growth Hormone, Human Pituitary, Iodination Grade, CAS 12629-01-5, is a single chain polypeptide hormone essential for growth of all tissues and plays a role in fat mobilization.

Synonym(s):

Growth Hormone, Human Pituitary, Iodination Grade, hGH, Somatotropin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥97% (weight)

form

lyophilized solid

specific activity

≥3.5 IU/mg (WHO 1st IRP)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.05% hFSH, hPRL, hLH, hTSH

pI 

4.9

solubility

0.01 M sodium bicarbonate: soluble

storage temp.

−20°C

General description

Native, human growth hormone from pituitary gland. Single chain polypeptide hormone essential for growth of all tissues. Increases fat mobilization. Immunopotency: ≥5 IU/mg (WHO 1st IRP).
Native, human growth hormone from pituitary gland. Single chain polypeptide hormone essential for the growth of all tissues. Increases fat mobilization. Secreted in a pulsatile manner.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

de Vos, A.M., et al. 1992. Science255, 306.
Brook, C.G.D., et al. 1988. J.Endocrinol.119, 179.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service