Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.55004

Sigma-Aldrich

Rink Amide AM resin (200-400 mesh)

Novabiochem®

Synonym(s):

4-(2′,4′-Dimethoxyphenyl-Fmoc-aminomethyl)-phenoxyacetamido-norleucylaminomethyl resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

General description

This support comprises the modified Rink amide linker attached via norleucine to aminomethylpolystyrene, and is an ideal tool for the Fmoc SPPS of peptide amides. Cleavage from this resin can be effected by a single step treatment with 95% TFA, providing peptide amides in high yields and purities.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Application


  • Second generation amphiphilic poly-lysine dendrons inhibit glioblastoma cell proliferation without toxicity for neurons or astrocytes: Uses Rink Amide AM resin with a 0.71mmol/g loading capacity for synthesizing amphiphilic dendritic amides. (Janiszewska et al., 2016).

  • The Role of Phosphoglycans in the Susceptibility of Leishmania mexicana to the Temporin Family of Anti-Microbial Peptides: Describes peptide synthesis on Rink Amide AM resin with a 0.79 mmol/g loading capacity. (Eggimann et al., 2015).

Linkage

Replaces: 01-64-0038

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.80 mmol/g
TNBS-Test: passes test
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.1 ml/g
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene-1 % DVB), 200 -400 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service