Skip to Content
MilliporeSigma

8.52003

Fmoc-Ala-OH

Novabiochem®

Synonym(s):

Fmoc-Ala-OH, N-α-Fmoc-L-alanine monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H17NO4
CAS Number:
Molecular Weight:
311.33
UNSPSC Code:
12352209
EC Index Number:
252-660-6
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

product line

Novabiochem®

assay

≥93.0% (acidimetric), ≥98% (TLC), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

140-150 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

SMILES string

N(C(C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16H,10H2,1H3,(H,19,22)(H,20,21)

InChI key

QWXZOFZKSQXPDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Standard building block for introduction of alanine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Application

  • Interface Passivation of Perovskite Solar Cells by Fmoc-Ala-OH Amino Acids: This study explores the effects of Fmoc-Ala-OH on perovskite solar cells, showing its potential to improve the efficiency and stability of these devices (Song et al., 2023).
  • Charge and Sequence Effects on the Self-Assembly and Subsequent Hydrogelation of Fmoc-depsipeptides: Discusses the role of Fmoc-Ala-OH in the self-assembly processes necessary for creating advanced materials, highlighting its utility in material science (Nguyen et al., 2014).
  • High-Quality Conjugated Polymers Achieving Ultra-Trace Detection of Cr2O72- in Agricultural Products: Illustrates the application of Fmoc-Ala-OH in the development of sensitive detection systems for environmental monitoring (Li et al., 2022).
  • A Liquid-Phase Continuous-Flow Peptide Synthesizer for Preparing C-terminal Free Peptides: Details how Fmoc-Ala-OH is used in innovative peptide synthesis technologies to streamline production processes in pharmaceutical applications (Otake et al., 2023).
  • Self-assembled Structures Formed by Fmoc Modified Aliphatic Amino Acids: Examines the use of Fmoc-Ala-OH in the formation of self-assembled structures, which are crucial for developing new materials with tailored properties (Gour et al., 2021).

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Ala-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 93.0 %
Water (K. F.): ≤ 6.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1006

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Song et al.
Cell reports, 30(8), 2699-2711 (2020-02-27)
The transcriptional corepressor complex CoREST is one of seven histone deacetylase complexes that regulate the genome through controlling chromatin acetylation. The CoREST complex is unique in containing both histone demethylase and deacetylase enzymes, LSD1 and HDAC1, held together by the

Global Trade Item Number

SKUGTIN
852003025004027269206918
852003500004027269221454
852003010004027269084431
852003100004027269179144
852003901504027269242800
852003902504027269217259
852003050004027269179120
852003901004027269221355
852003002504027269084424
852003200004027269232658
852003902004027269217273

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service