Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22336

Sigma-Aldrich

Thioglycolic acid

for synthesis

Synonym(s):

Thioglycolic acid, Mercaptoacetic acid, 2-Mercaptoethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O2S
CAS Number:
Molecular Weight:
92.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-677-4
NACRES:
NA.22

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

form

liquid

potency

73 mg/kg LD50, oral (Rat)

pH

1 (20 °C in H2O)

bp

220 °C/1013 hPa (decomposition)

mp

-16.5 °C

transition temp

flash point 131.5 °C

density

1.325 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C2H4O2S/c3-2(4)1-5/h5H,1H2,(H,3,4)

InChI key

CWERGRDVMFNCDR-UHFFFAOYSA-N

Application

  • Biocompatible Drug Delivery Films: Thioglycolic acid enhances the film-forming ability and bioadhesiveness of mucilaginous polysaccharides derived from Cassia uniflora seeds. This innovation is pivotal in drug delivery applications, offering controlled release and improved mucosal adhesion properties (Deore et al., 2024).
  • Surface Plasmon Resonance in Nanoparticle Research: The interfacial reactivity of gold nanoparticles can be inferred via surface plasmon resonance measurements, with thioglycolic acid playing a crucial role in modifying nanoparticle surfaces to enhance their reactive properties. This is significant for advancements in nanotechnology and material sciences (Romain et al., 2023).
  • Enhanced SERS for Virus Detection: Thioglycolic acid contributes to the fabrication of flexible electrospinning nano-fiber membranes that are pivotal in the detection of respiratory tract transmission viruses using Surface Enhanced Raman Scattering (SERS). This application is crucial in improving public health surveillance and response systems (Sun et al., 2024).

Analysis Note

Assay (iodometric): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 1.325 - 1.335
Identity (IR): passes test
The assay stated on the certificate of analysis refers to the date of examination. A gradual substance-specific content reduction, especially at higher temperatures, must be expected. The average content reduction at the required storage temperature of +2°c to +8°C is about 1% per month.

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service