Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.22196

Sigma-Aldrich

Chromium(0) hexacarbonyl

for synthesis

Synonym(s):

Chromium(0) hexacarbonyl, Chromiumcarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr(CO)6
CAS Number:
Molecular Weight:
220.06
MDL number:
UNSPSC Code:
12161603
EC Index Number:
235-852-4
NACRES:
NA.22

vapor pressure

0.35 hPa ( 20 °C)

Quality Level

form

crystals

mp

149-150 °C

density

1.77 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/6CO.Cr/c6*1-2;

InChI key

KOTQLLUQLXWWDK-UHFFFAOYSA-N

Application

  • Stable Salts of Chromium Hexacarbonyl Cation: Research on the stability and properties of chromium hexacarbonyl cation expands the understanding of its role in coordination chemistry, potentially influencing applications in catalysis and materials science (Himmel, Breher, Krossing).
  • Synthesis and Biological Evaluation: The use of chromium hexacarbonyl in the synthesis of organometallic complexes with biological activity, such as anticancer properties, suggests its utility in medicinal chemistry (Ha et al., 2023).
  • Catalysis for Fuel Cells: Employed in the synthesis of efficient nanowire cathode catalysts for proton exchange membrane fuel cells, demonstrating its application in green energy technologies (Liu et al., 2020).
  • Functionalization of 2D Materials: Used for the photochemical functionalization of gray arsenic, enabling applications in electronic and photonic devices, relevant for material science research (Sturala et al., 2019).
  • Dissociative Electron Attachment Studies: Investigated for its interactions with low-energy electrons, relevant in fields like focused electron beam induced deposition (FEBID), impacting nanofabrication technologies (Kopyra et al., 2017).

Analysis Note

Assay (ex Cr): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service