Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21111

Sigma-Aldrich

2-Thiazoline-2-thiol

for synthesis

Synonym(s):

2-Thiazoline-2-thiol, 2-Mercaptothiazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NS2
CAS Number:
Molecular Weight:
119.21
MDL number:
UNSPSC Code:
12352113
EC Index Number:
202-512-1
NACRES:
NA.22

Quality Level

form

powder

potency

300 mg/kg LD50, oral (Rat)

pH

6-7 (20 °C, 10 g/L in H2O)

mp

101-106 °C

storage temp.

2-30°C

InChI

1S/C3H5NS2/c5-3-4-1-2-6-3/h1-2H2,(H,4,5)

InChI key

WGJCBBASTRWVJL-UHFFFAOYSA-N

Application

  • Inducing apoptosis in melanoma cells: A study described a ruthenium(II) complex with 2-mercaptothiazoline ligand inducing selective cytotoxicity by causing DNA damage and apoptosis specifically in melanoma cells, highlighting its potential as a therapeutic agent in oncology (de Melo et al., 2024).
  • Antimicrobial and synergistic activities: Thiazoline derivatives, including compounds related to 2-mercaptothiazoline, were studied for their antimicrobial and synergistic effects with conventional antibiotics against multidrug-resistant Staphylococcus aureus, presenting a potential strategy for enhancing antibiotic efficacy (Khan et al., 2020).
  • Antimicrobial investigation of metal complexes: The antimicrobial properties of Cd(II) and Sn(II) complexes with 2-mercaptothiazoline were evaluated, underscoring the compound′s role in enhancing the bioactivity of metal-based therapeutics (Gaber et al., 2021).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service