Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20925

Sigma-Aldrich

2-Octanol

for synthesis

Synonym(s):

2-Octanol, Hexyl methyl carbinol, 2-Capryl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-667-0
NACRES:
NA.22
Assay:
≥98% (GC)
bp:
177-182 °C/1013 hPa
vapor pressure:
0.25 hPa ( 20 °C)

vapor pressure

0.25 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

oil

autoignition temp.

255-260 °C

potency

6100 mg/kg LD50, oral (Rat)

expl. lim.

0.80-7.40 % (v/v)

bp

177-182 °C/1013 hPa

mp

-38 °C

transition temp

flash point 71 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CCCCCC)C

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3

InChI key

SJWFXCIHNDVPSH-UHFFFAOYSA-N

Application

  • Development of a mixed solvent system for the efficient resolution of (R, S)-2-octanol catalyzed by magnetite-immobilized lipase: This study presents a novel solvent system enhancing the enantioselectivity and activity of lipase in the resolution of 2-octanol, a key intermediate in synthesizing chiral building blocks used in drug development (Liu et al., 2014).
  • Efficient Kinetic Resolution of (R,S)-2-Octanol Catalyzed by Magnetite-Immobilized Yarrowia lipolytica Lipase in Mixed Ionic Liquids: This research explores using ionic liquids with immobilized lipase for resolving 2-Octanol, crucial for producing enantiopure substances in pharmacological synthesis (Liu et al., 2014).
  • Effect of hydrazyl radical on the selectivity of γ-alumina with different sizes of pores for dehydration of 2-octanol through radical elimination mechanism: Investigates the interaction between hydrazyl radicals and 2-octanol on γ-alumina, relevant for understanding catalytic mechanisms in material science (Alidoust et al., 2023).
  • Process hazard evaluation for catalytic oxidation of 2-octanol with hydrogen peroxide using calorimetry techniques: Assesses the safety and kinetic parameters of 2-octanol oxidation, important for chemical process design in industrial and academic settings (Sun et al., 2019).
  • Enhancing the resolution of (R, S)-2-octanol catalyzed by magnetic cross-linked lipase aggregates using an alternating magnetic field: Describes a method to improve the enantioselective resolution of 2-Octanol using magnetic fields, potentially useful for scaling up chiral resolutions in pharmaceutical manufacturing (Liu et al., 2015).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.820 - 0.822
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service