Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20032

Sigma-Aldrich

2-Ethyl-1,3-hexanediol

(mixture of DL-and meso-form) for synthesis

Synonym(s):

2-Ethyl-1,3-hexanediol, Ethylhexylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
146.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-377-9
NACRES:
NA.22

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

340 °C (DIN 51794)

potency

5170 mg/kg LD50, oral (Rat)

refractive index

n22/589 1.4530

bp

241-249 °C/1013 hPa

mp

-40 °C

transition temp

flash point 136 °C

solubility

42 g/L

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H18O2/c1-3-5-8(10)7(4-2)6-9/h7-10H,3-6H2,1-2H3

InChI key

RWLALWYNXFYRGW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.940 - 0.942
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service