Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18801

Sigma-Aldrich

2,3-Butanediol

(mixture of meso- D- and L-form) for synthesis

Synonym(s):

2,3-Butanediol, 2,3-Butylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(OH)CH3
CAS Number:
Molecular Weight:
90.12
MDL number:
UNSPSC Code:
12352104
EC Index Number:
208-173-6
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

395 °C (DIN 51794)

potency

>5000 mg/kg LD50, oral (Rat)

expl. lim.

3.1-11.4 % (v/v)

pH

7 (20 °C in H2O, Aqueous solution)

bp

180-182 °C/1013 hPa

mp

18-20 °C

transition temp

flash point 96 °C (DIN 51758)

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O[C@H]([C@H](O)C)C

InChI

1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3/t3-,4+

InChI key

OWBTYPJTUOEWEK-ZXZARUISSA-N

Analysis Note

Assay isomers (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.000 - 1.005
Water (K. F.): ≤ 2.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service