Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.18710

Sigma-Aldrich

Urea

for synthesis

Synonym(s):

Carbamide, Carbonyl diamide, Diaminomethanone, Carbonyldiamine, Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-315-5
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

potency

8471 mg/kg LD50, oral (Rat)
8200 mg/kg LD50, skin (Rat)

pH

9 (20 °C, 100 g/L in H2O)

mp

132-135 °C (lit.)

solubility

1000 g/L

density

1.335 g/mL at 25 °C (lit.)

bulk density

720‑760 kg/m3

storage temp.

2-30°C

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Analysis Note

Assay (ex N): ≥ 99.0 %
Melting range (lower value): ≥ 132 °C
Melting range (upper value): ≤ 135 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service