Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.18370

Sigma-Aldrich

Imidazole-4,5-dicarboxylic acid

for synthesis

Synonym(s):

Imidazole-4,5-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O4
CAS Number:
Molecular Weight:
156.10
MDL number:
UNSPSC Code:
12352106
EC Index Number:
209-327-5
NACRES:
NA.22

Quality Level

form

powder

autoignition temp.

430 °C

pH

3.7 (20 °C, 0.5 g/L in H2O)

mp

270-275 °C

solubility

0.5 g/L

bulk density

650‑660 kg/m3

storage temp.

2-30°C

InChI

1S/C5H4N2O4/c8-4(9)2-3(5(10)11)7-1-6-2/h1H,(H,6,7)(H,8,9)(H,10,11)

InChI key

ZEVWQFWTGHFIDH-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service