Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18105

Sigma-Aldrich

1,1,3,3-Tetramethylbutyl isocyanide

for synthesis

Synonym(s):

1,1,3,3-Tetramethylbutyl isocyanide, Thiocarbamide, 1,1,3,3,-Tetramethylbutylisonitrile, Walborsky reagent, tert-Octylisonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2C(CH3)2NC
CAS Number:
Molecular Weight:
139.24
MDL number:
UNSPSC Code:
12352117
EC Index Number:
238-577-8
NACRES:
NA.22

Quality Level

Assay

≥96.0% (GC)

form

liquid

bp

55-56 °C/15 hPa

transition temp

flash point 51 °C

density

0.80 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+](#[C-])C(CC(C)(C)C)(C)C

InChI

1S/C9H17N/c1-8(2,3)7-9(4,5)10-6/h7H2,1-5H3

InChI key

YVPXQMYCTGCWBE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.799 - 0.801
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service