Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.14070

Sigma-Aldrich

Sebacic acid dinitrile

for synthesis

Synonym(s):

Sebacic acid dinitrile, Decanedinitrile, Decanedioic acid dinitrile, Sebaconitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CN(CH2)8CN
CAS Number:
Molecular Weight:
164.25
UNSPSC Code:
12352117
EC Index Number:
217-494-0
NACRES:
NA.22

Quality Level

Assay

≥94.5% (GC)

form

liquid

bp

199-204 °C/20 hPa

mp

7-8 °C

transition temp

flash point >93 °C

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H16N2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-8H2

InChI key

DFJYZCUIKPGCSG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 94.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.913 - 0.914
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service