Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08388

Sigma-Aldrich

1,2,4-Triazole

for synthesis

Synonym(s):

1,2,4-Triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3N3
CAS Number:
Molecular Weight:
69.07
MDL number:
UNSPSC Code:
12352005
EC Index Number:
206-022-9
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

solid

potency

1650 mg/kg LD50, oral (Rat)
3129-4200 mg/kg LD50, skin (Rabbit)

pH

8 (20 °C, 10 g/L in H2O)

bp

259-261 °C/1013 hPa

mp

119-122 °C

density

1.15 g/cm3 at 130 °C

bulk density

550 kg/m3

storage temp.

2-30°C

InChI

1S/C2H3N3/c1-3-2-5-4-1/h1-2H,(H,3,4,5)

InChI key

NSPMIYGKQJPBQR-UHFFFAOYSA-N

Application

  • Chemistry of 1, 2, 4-triazole: a review article: This comprehensive review explores the broad applications of 1,2,4-triazole in pharmaceuticals, demonstrating its role in enhancing antibacterial, antifungal, and antioxidant properties (Shneine et al., 2016).
  • Combination of 1, 2, 3-triazole and 1, 2, 4-triazole frameworks for new high-energy and low-sensitivity compounds: Discusses the synthesis of energetic materials combining 1,2,4-triazole with other high-energy frameworks, aiming to enhance performance and safety (Yao et al., 2021).
  • 1, 3, 4-Oxadiazole, 1, 3, 4-thiadiazole and 1, 2, 4-triazole derivatives as potential antibacterial agents: Examines derivatives of 1,2,4-triazole for potential use in antibacterial treatments, reflecting on their pharmacological importance (Othman et al., 2019).
  • Recent developments on 1, 2, 4-triazole nucleus in anticancer compounds: a review: Reviews the incorporation of the 1,2,4-triazole nucleus in various anticancer compounds, showcasing the therapeutic potential of this moiety (Kaur et al., 2016).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 117 °C
Melting range (upper value): ≤ 121 °C
Identity (IR): passes test

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service