Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08261

Sigma-Aldrich

Trifluoroacetic anhydride

for synthesis

Synonym(s):

Trifluoroacetic anhydride, Bis(trifluoroacetic) anhydride, TFAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO)2O
CAS Number:
Molecular Weight:
210.03
MDL number:
UNSPSC Code:
12352106
EC Index Number:
206-982-9
NACRES:
NA.22

vapor pressure

405 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

bp

40 °C/1013 hPa

mp

-65 °C

density

1.51 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4F6O3/c5-3(6,7)1(11)13-2(12)4(8,9)10

InChI key

QAEDZJGFFMLHHQ-UHFFFAOYSA-N

Application


  • Cascade Rearrangement: Nitro Group-Participating Syntheses of 1,2,5-Thiadiazoles and 1,2,4-Thiadiazolones.: This study explores the role of trifluoroacetic anhydride in the synthesis of thiadiazoles and thiadiazolones via a nitro group-participating cascade rearrangement, highlighting its utility in organic synthesis (Li et al., 2024).

  • Copper Loading-Controlled Selective Synthesis of 2,5-Bis(trifluoromethyl) and Monotrifluoromethyl-Substituted Oxazoles.: This research demonstrates the selective synthesis of trifluoromethyl-substituted oxazoles using trifluoroacetic anhydride, showcasing its versatility in controlling copper loading during reactions (Wu et al., 2024).

  • Wood Esterification by Fatty Acids Using Trifluoroacetic Anhydride as an Impelling Agent and Its Application for the Synthesis of a New Bioplastic.: The study presents a novel method for wood esterification using trifluoroacetic anhydride, facilitating the development of new bioplastic materials (Sejati et al., 2023).

  • Synthesis of alpha-Seleno Boronates via Diboration of Carbonyl Compounds.: This paper details the synthesis of alpha-seleno boronates using trifluoroacetic anhydride in the diboration of carbonyl compounds, emphasizing its reactivity and selectivity in chemical transformations (Paul et al., 2023).

  • Regioselective Synthesis of 3-Nitroindoles Under Non-Acidic and Non-Metallic Conditions.: The research highlights the use of trifluoroacetic anhydride in the regioselective synthesis of 3-nitroindoles, demonstrating its efficiency under non-acidic and non-metallic conditions (Zhang et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.509 - 1.512
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service