Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00608

Sigma-Aldrich

Propionic anhydride

for synthesis

Synonym(s):

Propionic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-638-2
NACRES:
NA.22

vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

285 °C

potency

2360 mg/kg LD50, oral (Rat)
10000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-9.5 % (v/v)

pH

2.5 (20 °C, 100 g/L in H2O, calculated on the free acid)

bp

167 °C/1013 hPa

mp

-45 °C

transition temp

flash point 63 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H10O3/c1-3-5(7)9-6(8)4-2/h3-4H2,1-2H3

InChI key

WYVAMUWZEOHJOQ-UHFFFAOYSA-N

Application

<ul>
<li><strong>Trace Amines Quantification:</strong> Propionic anhydride is used to derivatize the amino and hydroxyl groups for catecholamines and indolesin (de Bruyn et al., 2024).</li>
<li><strong>Asphaltene Oxidation Research:</strong> It is used in the oxidation of asphaltene and in the synthesis of peroxypropionic acid (Zhao et al., 2023).</li>
<li><strong>In Acylation Reactions:</strong> Used in the acylation of 1,2-methylenedioxybenzene (MDB) during the synthesis of 3,4-methylenedioxypropiophenone (MDP1P) and propionic acid.<br /> (Schiaroli et al., 2023).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.009 - 1.011
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

145.4 °F

Flash Point(C)

63 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service