Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69839-M

Sigma-Aldrich

T4 DNA Ligase - Novagen

Qualified under the most stringent cloning conditions

Synonym(s):

DNA Ligation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106310
NACRES:
NA.54

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice
wet ice

storage temp.

−20°C

General description

T4 DNA Ligase catalyzes the formation of phosphodiester bonds between 3′-hydroxyl and 5′-phosphate groups of adjacent DNA and RNA nucleotides in blunt-end or cohesive-end configurations. The enzyme joins RNA and DNA strands only in duplex molecules and does not join single-stranded nucleic acids. Novagen T4 DNA Ligase is rigorously tested in a blue-white cloning assay under conditions that maximize exposure of DNA ends to any contaminating nucleases. The enzyme is qualified for any ligation or cloning application.

Warning

Toxicity: Standard Handling

Unit Definition

4 Weiss units/µl; one Weiss unit is defined as the amount of enzyme required to catalyze the exchange of 1 nmole of ³²P from pyrophosphate into ATP as Norit®-absorbable material in 20 minutes at 37°C. 0.01 Weiss unit is the amount of enzyme required to ligate > 95% of 1 µg bacteriophage λ Hind III fragments at 16°C in 20 minutes.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Norit is a registered trademark of Norit N.V.

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service