Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681668

Sigma-Aldrich

Wnt Synergist, QS11

A cell-permeable purine compound that binds ARFGAPs (Kd = 364 and 620 nM for AMAP1 and ARFGAP1, respectively) and acts as a broad specificity ARFGAPs inhibitor (GTPase activating proteins of ADP-ribosylation factor).

Synonym(s):

Wnt Synergist, QS11, QS11, Wnt Pathway Activator II, Wnt Pathway Activator II, QS11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H33N5O2
CAS Number:
Molecular Weight:
567.68
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale amber

solubility

ethanol: 20 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C36H33N5O2/c42-23-31(20-25-8-3-1-4-9-25)38-34-33-35(40-36(39-34)43-32-19-18-28-12-7-13-30(28)21-32)41(24-37-33)22-26-14-16-29(17-15-26)27-10-5-2-6-11-27/h1-6,8-11,14-19,21,24,31,42H,7,12-13,20,22-23H2,(H,38,39,40)/t31-/m0/s1

InChI key

DOKZLKDGUQWMSX-HKBQPEDESA-N

General description

A cell-permeable purine compound that binds ARFGAPs (Kd = 364 and 620 nM for AMAP1 and ARFGAP1, respectively) and acts as a broad specificity ARFGAPs inhibitor (GTPase activating proteins of ADP-ribosylation factor). Shown to synergize with Wnt-3a and specifically activate the canonical Wnt/β-catenin signaling pathway both in vitro (2-, 40-, and 200-fold increase of basal transcription activity by QS11, Wnt-3a, and QS11/Wnt-3a, respectively, in a Super(8X)TOPFlash reporter assay using HEK293 cells) and in vivo (in the induction of Xenopus embryonic axis duplication). Also reported to block the migration of AMAP1-overexpressing MDA-MB-231 cells (86% inhibition at 2.5 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, Q., et al. 2007. Proc. Natl. Acad. Sci. USA104, 7444.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service