Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

506172

Sigma-Aldrich

p38 MAP Kinase Inhibitor X, BIRB 796

The p38 MAP Kinase Inhibitor X, BIRB 796, also referenced under CAS 285983-48-4, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

p38 MAP Kinase Inhibitor X, BIRB 796, Doramapimod, BIRB796, 1-(5- tert-Butyl-2- p-tolyl-2H-pyrazol-3-yl)-3-[4-(2-morpholin-4-yl-ethoxy)naphthalen-1-yl]urea, JNK Inhibitor XVII, Doramapimod, BIRB796, 1-(5-tert-Butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-[4-(2-morpholin-4-yl-ethoxy)naphthalen-1-yl]urea, JNK Inhibitor XVII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H37N5O3
CAS Number:
Molecular Weight:
527.66
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC1=CC=C(N2C(NC(NC3=CC=C(OCCN4CCOCC4)C5=C3C=CC=C5)=O)=CC(C(C)(C)C)=N2)C=C1

InChI

1S/C31H37N5O3/c1-22-9-11-23(12-10-22)36-29(21-28(34-36)31(2,3)4)33-30(37)32-26-13-14-27(25-8-6-5-7-24(25)26)39-20-17-35-15-18-38-19-16-35/h5-14,21H,15-20H2,1-4H3,(H2,32,33,37)

InChI key

MVCOAUNKQVWQHZ-UHFFFAOYSA-N

General description

A cell-permeable pyrazolo-napthalenyl urea compound that acts as a highly potent, slowly binding (IC50 = 8 and 97 nM, respectively, against human p38α, respectively, with or without 2 h preincubation), but high affinity (t1/2 of dissociation from human p38α = 23 h), p38-selective inhibitor by simultaneously targeting the ATP-binding site and an adjacent DFG-out conformation-specific allosteric cavity. It inhibits JNK2α2 and c-Raf-1 only at much higher concentrations (IC50 = 98 nM and 1.4 µM, respectively) and displays little or no activity toward 10 other kinases (IC50 >20 µM). Shown to inhibit LPS-induced TNF-α production both in vitro (IC50 = 18 and 780 nM using THP-1 cells and in human whole blood, respectively) and in mice in vivo (by 84%; 30 mg/kg p.o.).
A cell-permeable pyrazolo-napthalenyl urea compound that acts as a highly potent, slowly binding (IC50 = 8 and 97 nM, respectively, against human p38α, respectively, with or without 2 h preincubation), but high affinity (t1/2 of dissociation from human p38α = 23 h), p38-selective inhibitor by simultaneously targeting the ATP-binding site and an adjacent DFG-out conformation-specific allosteric cavity. It inhibits JNK2α2 and c-Raf-1 only at much higher concentrations (IC50 = 98 nM and 1.4 µM, respectively) and displays little or no activity toward 10 other kinases (IC50 >20 µM). Shown to inhibit LPS-induced TNF-α production in cultures in vitro (IC50 = 18 and 780 nM using THP-1 cells and in human whole blood, respectively) and in mice in vivo (by 84% and 63%, respectively, with oral dosage of 30 mg/kg and 10 mg/kg).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Liu. Y. and Gray, N.S., 2006. Nat. Chem. Biol.2, 358.
Regan, J., et al. 2003. J. Med. Chem.46, 4676.
Pargellis, C., et al. 2002. Nat. Struct. Biol.9, 268.
Regan, J., et al. 2002. J. Med. Chem.45, 2994.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Woo Joo Lee et al.
Journal of cellular physiology, 237(7), 2943-2960 (2022-05-03)
Growing evidence indicates that long intergenic noncoding RNAs play an important role in cancer progression by affecting gene regulation at the transcriptional and posttranscriptional levels. Recent studies have shown that long intergenic noncoding RNA functions as a competitive endogenous RNA
Shah Md Toufiqur Rahman et al.
The Journal of biological chemistry, 295(25), 8494-8504 (2020-05-07)
The selective pressure imposed by extrinsic death signals and stressors adds to the challenge of isolating and interpreting the roles of proteins in stress-activated signaling networks. By expressing a kinase with activating mutations and a caged lysine blocking the active
Adil Mohamed et al.
Journal of virology, 97(2), e0000923-e0000923 (2023-02-07)
Mammalian orthoreovirus serotype 3 Dearing is an oncolytic virus currently undergoing multiple clinical trials as a potential cancer therapy. Previous clinical trials have emphasized the importance of prescreening patients for prognostic markers to improve therapeutic success. However, only generic cancer
Munmun Chakraborty et al.
Cells, 10(12) (2021-12-25)
The lack of an animal model or an in vitro model limits experimental options for studying temporal molecular events in pseudoexfoliation syndrome (PXF), an age related fibrillopathy causing trabecular meshwork damage and glaucoma. Our goal was to create a workable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service