Skip to Content
MilliporeSigma

5.00862

SAFC

Vitamin E

water-miscible 1 ml = 500 mg all-rac-α-tocopheryl acetate for oral use, EMPROVE® ESSENTIAL, USP

Pharma Manufacturing

Synonym(s):

(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
94012
MDL number:
UNSPSC Code:
12352401

Agency

USP

Quality Level

product line

EMPROVE® ESSENTIAL

form

liquid

pH

6.5-7.5 (20 °C, 100 g/L in H2O)

density

0.950 g/mL at 20 °C (lit.)
1.01 g/cm3 at 20 °C

application(s)

pharmaceutical

storage temp.

15-25°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3

InChI key

GVJHHUAWPYXKBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Vitamin E protects cell against free radicals. It is used in skin and hair care products, as well as in dietary supplements. Vitamin E water-miscible 1 ml = 500 mg DL-α-tocopherol acetate for oral use, Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service