Skip to Content
MilliporeSigma
All Photos(1)

Documents

454126

Sigma-Aldrich

Methotrexate

≥98% (HPLC), solid, thymidylate synthetase inhibitor, Calbiochem®

Synonym(s):

Methotrexate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N8O5 · 3H2O
CAS Number:
Molecular Weight:
508.49
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Methotrexate,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/t13-/m0/s1

InChI key

FBOZXECLQNJBKD-ZDUSSCGKSA-N

General description

Inhibits thymidylate synthetase, is a nonselective de novo purine synthesis inhibitor and has a significant toxicity profile including hepatotoxicity, pneumonitis and bone marrow suppression. Potent folic acid antagonist. Induces apoptosis in HL-60 human leukemia cells. Also useful as an anti-tumor agent.

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kaufman, S.H., et al. 1993. Cancer Res.53, 3976.
Takasuga, A., et al. 1992. J. Biochem.112, 652.
Hirata, S., et al. 1989. Arthritis Rheum. 32, 1065.
Jolivet, J., et al. 1983. New Engl. J. Med. 309, 1094.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 1B - STOT RE 1

Target Organs

Liver,Bone marrow

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ofer Shoshani et al.
Nature, 591(7848), 137-141 (2020-12-29)
Focal chromosomal amplification contributes to the initiation of cancer by mediating overexpression of oncogenes1-3, and to the development of cancer therapy resistance by increasing the expression of genes whose action diminishes the efficacy of anti-cancer drugs. Here we used whole-genome sequencing
Lukas Westermann et al.
Scientific reports, 12(1), 18211-18211 (2022-10-29)
Genome editing tools such as CRISPR/Cas9 enable the rapid and precise manipulation of genomes. CRISPR-based genome editing has greatly simplified the study of gene function in cell lines, but its widespread use has also highlighted challenges of reproducibility. Phenotypic variability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service