Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43-040

Millipore

MILLIPLEX MAP Lysis buffer for Multiplexing

Synonym(s):

Buffer for MILLIPLEX, Lysis Buffer for Multiplexing, Multiplex Lysis Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.25

form

solution

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

Application

Research Category
All
This MILLIPLEX MAP Lysis Buffer is a buffer used for multiplex Cell Signaling bead-based kits.

Storage and Stability

stable 1 year at 4°C from date of shipment

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rasha Al-Attar et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 242, 110653-110653 (2020-01-12)
Naked mole rats (NMRs, Heterocephalus glaber) are among the most hypoxia-tolerant mammals known. They can reduce their metabolic rate (>85%) under severe hypoxia, remain moderately active and recover with no obvious signs of damage. Hence, NMRs are an excellent model
Aakriti Gupta et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 243-244, 110436-110436 (2020-04-05)
The wood frog (Rana sylvatica) is a remarkable species. These frogs can endure prolonged oxygen deprivation as well as dehydration to ~60% of total body water lost and, combining these two abilities, they survive whole body freezing for weeks at
Cheng-Wei Wu et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 246-247, 110461-110461 (2020-06-05)
In arid conditions, the African Clawed frog Xenopus laevis enters a state of estivation dormancy as an adaptive survival strategy. Under estivation, X. laevis experience severe dehydration stress as 25-35% of total body water is lost. Dehydration in X. laevis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service