Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

371955-M

Millipore

H-7, Dihydrochloride - CAS 108930-17-2 - Calbiochem

A broad-based, cell-permeable, reversible, ATP-competitive serine/threonine kinase inhibitor.

Synonym(s):

H-7, Dihydrochloride - CAS 108930-17-2 - Calbiochem, 1-(5-Isoquinolinesulfonyl)-2-methylpiperazine, 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N3O2S · 2HCl
CAS Number:
Molecular Weight:
364.29
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (TLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

water: 20 mg/mL

storage temp.

2-8°C

InChI

1S/C14H17N3O2S.2ClH/c1-11-9-16-7-8-17(11)20(18,19)14-4-2-3-12-10-15-6-5-13(12)14;;/h2-6,10-11,16H,7-9H2,1H3;2*1H

InChI key

OARGPFMFRLLKPF-UHFFFAOYSA-N

General description

A broad-based, cell-permeable, reversible, ATP-competitive serine-threonine kinase inhibitor. Potent inhibitor of myosin light chain kinase (Ki = 97 µM), protein kinase A (Ki = 3.0 µM), protein kinase C (Ki = 6 µM), and protein kinase G (Ki = 5.8 µM). Induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells. Also inhibits telomerase activity in quercetin-, H-89-, or herbimycin A-treated NPC-076 cells.
A broad-based, cell-permeable, reversible, ATP-competitive serine/threonine kinase inhibitor. Inhibits protein kinase A (Ki = 3.0 µM), myosin light chain kinase (Ki = 97 µM), protein kinase C (Ki = 6.0 µM), and protein kinase G (Ki = 5.8 µM). Induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells. Also inhibits telomerase activity in quercetin-, H-89-, or herbimycin A-treated NPC-076 cells.

Biochem/physiol Actions

Primary Target
MLCK
Target Ki: 3.0 µM, 97 µM, 6.0 µM, 5.8 µM against protein kinase A , myosin light chain kinase, protein kinase C , and protein kinase G, respectively

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Aqueous stock solutions are stable for up to 6 months at 4°C.

Other Notes

Ku, W.-C., et al. 1997. Biochem. Biophys. Res. Commun. 241, 730.
Jarvis, W.D., et al. 1994. Cancer Res.54, 1707.
Bergstrand, H., et al. 1992. J. Pharmacol. Exp. Therap.263, 1334.
Gimond, C., et al. 1992. Exp. Cell Res.203, 365.
Quick, J., et al. 1992. Biochem. Biophys. Res. Commun.187, 657.
Bouli, N.M., and Davis, M. 1990. Brain Res.525, 198.
Takahashi, I., et al. 1990. J. Pharmacol. Exp. Ther.255, 1218.
Schachtele, C., et al. 1988. Biochem. Biophys.Res. Commun.151, 542.
Kawamoto, S., and Hidaka, H. 1984. Biochem. Biophys. Res. Commun.125, 258.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service