Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341274-M

Sigma-Aldrich

Factor H, Human

Factor H, Human, is a single-chain multi-functional glycoprotein that regulates formation and function of complement C3 and C5 convertase enzymes.

Synonym(s):

β 1H Globulin, β1H Globulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, Factor I, Factor B, Factor P, C3, C4, or albumin, trace (Ouchterlony)

storage temp.

−70°C

General description

Native factor H from human serum. Single-chain glycoprotein present in normal human serum at about 500 µg/ml. Regulates formation and function of complement C3 and C5 convertase enzymes. A C3b-binding protein, not an enzyme. Regulatory activity is attributed to its ability to recognize and bind to C3b fragments.
Native factor H from human serum. Single-chain multi-functional glycoprotein present in normal human serum at about 500 µg/ml. Regulates formation and function of complement C3 and C5 convertase enzymes. A C3b-binding protein that does not exhibit enzymatic activity. Regulatory activity is attributed to its ability to recognize and bind to C3b fragments.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Zipfel, P.F., et al. 1999. Immunopharmacology 42, 53.
Pangburn, M.K. and Muller-Eberhard, H.J. 1984. Springer Semin. Immunopathol. 7, 163.
Fearon, D.T. and Austen, K.F. 1977. Proc. Natl. Acad. Sci. USA 74, 1683.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service