Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203780

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl-α-D-galactopyranoside

Synonym(s):

5-Bromo-4-chloro-3-indolyl-α-D-galactopyranoside, α-X-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.41

Assay

≥98% (TLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

2% DMF: soluble, clear

shipped in

ambient

storage temp.

−20°C

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14+/m1/s1

InChI key

OPIFSICVWOWJMJ-YGEXGZRRSA-N

General description

A chromogenic substrate for α-galactosidase. Useful for histochemical studies.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gossrau, R. 1990. Folia Histochem. Cytobiol. 28, 129.
Gossrau, R., and Lojda, Z. 1989. Acta Histochem. 85, 213.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service