Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

199001-M

Sigma-Aldrich

Benzamidine, Hydrochloride

Inhibitor of trypsin and trypsin-like enzymes.

Synonym(s):

Benzamidine, Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2 · xHCl
CAS Number:
Molecular Weight:
120.15 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: >15 mg/mL

storage temp.

10-30°C

InChI

1S/C7H8N2.ClH/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H3,8,9);1H

InChI key

LZCZIHQBSCVGRD-UHFFFAOYSA-N

General description

Inhibitor of trypsin and trypsin-like enzymes. Inhibits Factor VII autoactivation.

Biochem/physiol Actions

Primary Target
trypsin and trypsin-like enzymes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kam, C.M., et al. 1995. Arch. Biochem. Biophys. 316, 808.
Brandstetter, H., et al. 1992. J. Mol. Biol.226, 1085.
Nishimura, Y., et al. 1992. Jpn. J. Cancer Res.83, 723.
Reivinen, J., et al. 1992. Kidney Int. 42, 624.
Sturzebecher, J., et al. 1992. Biol. Chem. Hoppe Seyler 373, 1025.
Yamamoto, M., et al. 1992. J. Biol. Chem.267, 19089.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

YeastBuster™ Protein Extraction Reagent for Fast, Efficient Extraction of Proteins from Yeast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service