Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.94963

Supelco

Chromium(VI) oxide

EMPLURA®

Synonym(s):

Chromium(VI) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
MDL number:
UNSPSC Code:
12352303

product line

EMPLURA®

storage temp.

room temp

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service