Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93633

Supelco

Calcium chloride dihydrate

EMPLURA®

Synonym(s):

Calcium chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
MDL number:
UNSPSC Code:
12352302

grade

laboratory grade

vapor pressure

0.01 hPa ( at 20 °C)

product line

EMPLURA®

form

solid

pH

4.5-9.2 (at 20 °C, 50 g/L)

mp

176 °C (dec.) (lit.)

solubility

water: 745 g/L at at 20 °C (anhydrous)

density

1.85 g/cm3 at at 20 °C

storage temp.

room temp

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

<ul>
<li><strong>Understanding the core-shell interactions in macrocapsules of organic phase change materials and polysaccharide shell: </strong>Investigates the thermal properties and structural integrity of phase change materials encapsulated in polysaccharide shells, utilizing barium chloride dihydrate for stability testing in energy storage applications (Reddy VJ et al., 2022).</li>
<li><strong>Synthesis of a novel monofilament bioabsorbable suture for biomedical applications: </strong>Focuses on the development of a bioabsorbable suture material enhanced with barium chloride dihydrate, which improves the mechanical properties and usability in surgical procedures, contributing to the advancement of medical materials (de la Harpe KM et al., 2022).</li>
</ul>

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service