Skip to Content
MilliporeSigma
All Photos(1)

Documents

16-236

Sigma-Aldrich

Anti-phospho-erbB-2/HER-2 (Tyr1248) Antibody, Alexa Fluor 555

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr1248)

Related Categories

Specificity

Predicted to cross-react with rat based on conservation of immunogenic sequence
activated form of erbB2/HER-2 oncoprotein

Immunogen

Synthetic phosphopeptide, residues 1243-1255, corresponding to a major autophosphorylation (Tyr1248) site of human erbB2/HER-2 (C-AENPEpYLGLDVPV)

Application

Anti-phospho-erbB-2/HER-2 (Tyr1248) Antibody, Alexa Fluor 555 is an antibody against phospho-erbB-2/HER-2 (Tyr1248) for use in FC & IC.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Kinases & Phosphatases

Quality

Routinely evaluated by immunostaining of EGF treated A431 cells

Physical form

ImmunoAffinity Purified

Storage and Stability

1 year at 4°C from date of shipment

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service