Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46751

Millipore

Readyplate XLD Agar acc ISO 6579

For isolation and differentiation of Salmonella, box of 20 plates, plate diam. 90 mm

Synonym(s):

Xylose Lysine Deoxycholate Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Agency

APHA
ISO 19250
ISO 6579

Quality Level

form

solid

feature

ready-to-use

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

storage condition

(Store dry and tightly closed)

plate diam.

90 mm

color

red

application(s)

food and beverages
industrial qc
microbiology

storage temp.

15-25°C

suitability

Salmonella spp.

General description

XLD agar is a selective agar that enables the identification of Salmonella species over other bacteria. This medium relies on three indicator systems:
  1. Degradation of xylose, lactose, and sucrose to acid causes phenol red to change its color to yellow.
  2. Production of hydrogen sulfide is indicated by thiosulfate and iron (III) salt, which react to form a precipitate of black iron sulfide in the colonies.
  3. The appearance of a purple coloration around the colonies due to an increase in pH indicates bacteria that decarboxylate lysine to cadaverine.
These reactions can proceed simultaneously or successively and may cause the pH indicator to exhibit various shades of color, or it may change its color from yellow to red on prolonged incubation. The culture medium is moderately selective thus, the growth of unwanted organisms is suppressed, while sought bacteria can be tentatively grouped by reading the net effect of carbohydrate dissimilation, lysine decarboxylation, and the formation of hydrogen sulfide leading to black colonies. Agar is the solidifying agent.

Application

ReadyPlate XLD Agar plates are suitable for isolation and differentiation of Salmonella from food and animal feed, water, and other materials.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service