Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46702

Millipore

Sabouraud Dextrose Agar

Lecithin, Tween®, Histidine, Thiosulfate, lid, ICR plus; lockable, plate diam. 90 mm, sterile; γ-irradiated, triple packed of 20 and 120 plates

Synonym(s):

SDA, SDA settle plates, SDA with neutralizers, Sabouraud Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate (ready-to-use)

feature

lid ICR plus; lockable
ready-to-use

composition

Histidine
Lecithin
Thiosulfate
Tween®

packaging

triple packed of 20 and 120 plates

plate diam.

90 mm

color

clear yellowish Medium

pH

5.6 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

molds
yeasts

General description

Sabouraud Dextrose Agar + LTHTh - ICR in 90 mm settle plates (lockable) is a selective media for the determination of the total count of yeasts and molds in air via active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms for sterility testing. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favours fungal growth and inhibits contaminating bacteria from samples. Lecithin, polysorbate 80 (Tween®), histidine and sodium thiosulfate are neutralizers to inactivate residues of active agents such as Aldehyde, Biguanides, Oxidizing compounds, Phenolic compounds and quaternary ammonium compounds. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport after sampling without the risk of losing the lid as well as for aerobic incubation the plates should be locked in the “CLOSED”-position (turn the lid clockwise) and for anaerobic or microaerophilic incubation in the “VENT” position (turn the lid anti-clockwise)

Application

Sabouraud Dextrose Agar (SDA) is a selective medium for cultivation and isolation of yeasts and molds. The media finds its application in environmental monitoring by determining the yeast and mold count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.

Storage and Stability

  • The product can be used for sampling until the expiry date if stored upright, protected from light, and properly sealed at +15 °C to +25 °C.
  • Condensation can be prevented by avoiding quick temperature shifts and mechanical stress.
  • The testing procedures described on the CoA can be started up to the expiry date printed on the label.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service