Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46429

Millipore

Tryptic Soy Agar - Ready-to-use Bottles & Tubes

tube capacity 25 mL, tube filling volume 18 mL, closure type, blue screw cap with septum, box of 20 or 100 tubes

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, TSA, Tryptic Soy Agar, Tryptone Soya Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106204
NACRES:
NA.74

Agency

EP 2.6.13
JP 4.05
USP 62

Quality Level

description

For microbiological examination of non-sterile products

sterility

sterile

form

liquid (ready-to-use)

shelf life

12 mo.

feature

closure type blue screw cap with septum
ready-to-use

packaging

box of 20 or 100 tubes
single packed of (18 ml in 25 ml tube with blue cap (20 or 100 tubes per box))

technique(s)

direct inoculation: suitable
sterile filtration: suitable

tube capacity

25 mL

tube filling volume

18 mL

color

clear yellowish

pH

7.3

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

General description

Tryptic Soy Agar is a complex medium for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The several peptones in the medium provide amino acids and polypeptides to ensure the growth of a variety of microorganisms. Sodium chloride maintains osmotic equilibrium in the medium.

Application

Tryptic Soy Agar is suitable for the cultivation and isolation of fastidious bacteria, yeasts, and fungi. The media is suitable for a large range of applications, including cultivation, maintenance of stock culture, and total aerobic microbial count. Its applications also include phage and colicin typing, X- and V-factor tests, and geno- and electrotyping (e.g. pulsed-field gel electrophoresis). TSA is applied especially for the determination of bacterial contamination of cosmetics, drinking water, and wastewater.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service