Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46375

Millipore

Tryptic Soy Agar - Ready-to-use Bottles & Tubes

bottle capacity 500 mL, bottle filling volume 400 mL, closure type, blue screw cap with 3 loci, box of 6 bottles

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, Tryptone Soya Agar, TSA, Tryptic Soy Agar EP+USP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

description

For cultivation & isolation of fastidious bacteria, yeasts, and fungi

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type blue screw cap with 3 loci
ready-to-use

packaging

box of 6 bottles
single packed of (400 ml in 500 ml bottle with blue screw cap and 3 loci (6 bottles per box))

bottle capacity

500 mL

bottle filling volume

400 mL

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar (TSA) is a complex growth medium used for the cultivation of a wide variety of microorganisms. Peptones in the medium provide amino acids and polypeptides to ensure the growth of microorganisms, while sodium chloride maintains the osmotic equilibrium. This universal agar displays a wide application range, including phage and colicin typing, X- and V-factor tests, and geno- and electrotyping (e.g. pulsed-field gel electrophoresis). TSA in the bottle can be used to prepare plates for surface spread or pour plate method.

Application

Tryptic Soy Agar is used for the cultivation and isolation of fastidious bacteria, yeasts, and fungi. It has a wide range of applications, including cultivation, maintenance of stock culture, and total aerobic microbial count. TSA is mainly used to determine bacterial contamination in cosmetics, drinking water, and wastewater.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Our media are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service