Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46075

Millipore

HEIMPLATE R2A Agar

Heimplate, JP, EP, ICR Settle Plate for long Incubation, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

Reasoner´s 2A agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP
JP

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use
ICR Settle Plate for long Incubation

packaging

box of 20-120 plates

manufacturer/tradename

Heimplate

technique(s)

microbiological culture: suitable

plate diam.

90 mm

plate filling volume

30 mL

pH

7.2 ( in H2O)

application(s)

air monitoring
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

heterotrophic bacteria

General description

R2A Agar (Reasoner´s 2A agar) is designed to promote the growth of micro-organisms that grow in low nutrient environments, such as water. The reduced amount of nutrients in the medium allows the growth of aerobic, heterotrophic micro-organisms, which are adapted to low nutrient content. Each settle plate is of 90 mm in diameter, and the packs are single-bagged in transparent, hydrogen peroxide impermeable sleeves (non-irradiated).

Application

R2A Agar plates are designed for the determination of the total microbial count in water for injections in bulk, highly purified water, and purified water in bulk.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service