Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46052

Millipore

Sabouraud Dextrose Agar

Thiosulfate, Lecithin, Tween®, Histidine, plate diam. 90 mm , Settle plate for Long Incubation, sterile; aseptically filled, single packed of 20 and 120 plates

Synonym(s):

ICR settle plate, SDA, SDA with neutralizer, Sabouraud Dextrose Agar with Lecithin, Tween, Histidine, and Thiosulfate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; aseptically filled

form

plate (ready-to-use)

feature

ready-to-use

composition

Histidine
Lecithin
Thiosulfate
Tween®

packaging

single packed of 20 and 120 plates

plate diam.

90 mm , Settle plate for Long Incubation

color

clear yellowish Medium

pH

5.6 ( in H2O)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

15-25°C

suitability

molds
yeasts

General description

Sabouraud Dextrose Agar + LTHTh in 90 mm settle plate with 30 mL filling volume for long incubation is designed for the detection of yeasts and molds in the air via active or passive air monitoring as well as fingerprints of personnel in controlled environments in the presence of disinfectant residues.
Sabouraud Dextrose Medium contains digests of animal tissues (peptones), which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favors fungal growth and inhibits contaminating bacteria from samples. Additionally, the medium is supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, histidine, and Sodium thiosulphate to inactivate disinfectant residues.

Application

Sabouraud Dextrose Agar + LTHTh settle plate is designed for the detection of the total count of yeasts and molds in air and from personnel in less critical environments.

Other Notes

  • The product can be used for sampling until the expiry date if stored upright, protected from light, andproperly sealed at +15 °C to +25 °C.
  • Condensation can be prevented by avoiding quick temperature shifts and mechanical stress.
  • The testing procedures as described on the CoA can be started up to the expiry date printed on the label.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Settle plates for isolators and critical cleanrooms

Settle plates for isolators and critical cleanrooms

Settle plates for isolators and critical cleanrooms

Settle plates for isolators and critical cleanrooms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service