Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-519-M

Sigma-Aldrich

Cdk6/Cyclin D3 Protein, active, 10 µg

Active, N-terminal, His6-tagged full length human Cdk6 complexed with N-terminal GST-tagged full-length human Cyclin D3, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

mol wt

Mw 38 kDa (cdk6)
Mw 59 kDa (cyclin D3)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... CDK6(1021)

General description

N-terminal, His6-tagged fulllength human cdk6 complexed with N-terminal GSTtagged full-length human cyclin D3
Product Source: expressed in Sf21 cells

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Protein Target: CDK6/cyclinD3
Target Sub-Family: CMGC

Packaging

Also available in 250μg size as catalog # 14-519M - Call for pricing and availability.

Quality

routinely evaluated by phosphorylation of histone H1

Physical form

Glutathione-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service