Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-466-M

Sigma-Aldrich

Insulin Receptor Protein, active, 10 µg

Active, recombinant human Insulin Receptor residues 1005-1310, containing an N-terminal His6-tag, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 36.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... INSR(3643)

General description

Product Source: Expressed by baculovirus in Sf21 insect cells
Recombinant human Insulin receptor residues 1005-1310, containing an Nterminal His6-tag

Biochem/physiol Actions

Protein Target: IR
Target Sub-Family: TK

Quality

routinely evaluated by phosphorylation of Axl-tide

Physical form

Ni2+/NTA-agarose

Storage and Stability

1year at -70C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service