Skip to Content
MilliporeSigma

1.28201

SAFC

Sodium acetate anhydrous

EMPROVE® ESSENTIAL, ChP, USP, FCC, E 262

Pharma Manufacturing

Synonym(s):

Sodium acetate, Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
MDL number:
UNSPSC Code:
51191566
EC Index Number:
204-823-8

grade

anhydrous

Quality Level

Agency

ChP
USP

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

1112 °F

mol wt

82.03 g/mol

impurities

bacteria content, low

pH

7.5-9.2 (20 °C, 30 g/L in H2O)

pKa 

4.76 (acetic acid)
4.76

bp

>400 °C (decomposition)

mp

>300 °C (dec.) (lit.)
324 °C (decomposition)

transition temp

flash point >250 °C

density

1.52 g/cm3 at 20 °C

application(s)

liquid formulation
liquid formulation
pharmaceutical

storage temp.

15-25°C

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium acetate anhydrous is an acetate buffer system component commonly used in biomolecule downstream chromatography steps as well as in liquid formulation.

Packaging

1282010003//3X1kg - plastic bottle
1282011001//1kg - plastic bottle
1282018026//25kg - plastic drum

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service