Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.15118

Millipore

Ethylene glycol monomethyl ether

for amino acid analysis

Synonym(s):

2-Methoxyethanol, Methyl glycol, Ethylene glycol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1731074
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-713-7
NACRES:
NA.05
Assay:
≥99.5% (GC)
bp:
124-125 °C (lit.)
vapor pressure:
6.17 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.62 (vs air)

Quality Level

vapor pressure

6.17 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

548 °F

expl. lim.

24.5 %

refractive index

n20/D 1.402 (lit.)

pH

7 (20 °C)

kinematic viscosity

1.6 cSt(20 °C)

bp

124-125 °C (lit.)

mp

−85 °C (lit.)

transition temp

flash point 37 °C

density

0.965 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

COCCO

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for amino acid analysis

Application

Ethylene glycol monomethyl ether for amino acid analysis. CAS 109-86-4, EC Number 203-713-7, chemical formula CH₃OCH₂CH₂OH.

Specifications

Saturation Concentration in Air: 33 g/m3 (20 °C)

Analysis Note

Purity (GC)≥ 99.5 %Identity (IR)conformsFree acid (as CH₃COOH)≤ 0.005 %Peroxide (as H₂O₂)≤ 0.001 %Evaporation residue≤ 0.003 %Water≤ 0.1 %Suitability for amino acid analysisconformsThe product is stabilized with 2,6-Di-tert-butyl-4-methylphenol (BHT).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service