Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.11714

Millipore

3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide

for biochemistry

Synonym(s):

3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide, Thiazolyl blue, MTT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16BrN5S
CAS Number:
Molecular Weight:
414.32
MDL number:
UNSPSC Code:
12352107
EC Index Number:
206-069-5
NACRES:
NA.21

Quality Level

form

solid

mp

195 °C

solubility

10 g/L

storage temp.

2-30°C

InChI

1S/C18H16N5S.BrH/c1-13-14(2)24-18(19-13)23-21-17(15-9-5-3-6-10-15)20-22(23)16-11-7-4-8-12-16;/h3-12H,1-2H3;1H/q+1;/p-1

InChI key

AZKSAVLVSZKNRD-UHFFFAOYSA-M

Application

<ul>
<li><strong>Anal Biochem, 2017</strong>: Comparison of MTT and Neutral Red assays to assess cell viability in the presence of copper compounds, highlighting the versatility and sensitivity of the MTT assay in cytotoxicity testing (Gomez Perez et al., 2017).</li>
<li><strong>Indian J Pharmacol, 2016</strong>: Evaluation of the cytotoxic potential of various Indian fruit peels on liver cancer cells (HepG2) using MTT assay, providing insights into natural product research and potential anticancer properties (Garg et al., 2016).</li>
<li><strong>ACS Appl Mater Interfaces, 2015</strong>: Application in the real-time evaluation of live cancer cells, combining surface plasmon resonance and electrochemical methodologies with MTT assays for enhanced accuracy in viability testing (Wu et al., 2015).</li>
</ul>

Analysis Note

Assay (argentometric): ≥ 98
Identity (IR-spectrum): passes test
Spec. Absorptivity A 1%/1cm (242 nm; 0.001 %; 1 cm; methanol): 480 - 530
Heavy metals (as Pb): ≤ 0.001 %
Sulfated ash: ≤ 0.5 %

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service