Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09676

Supelco

2′,7′-Dichlorofluorescein

fluorenscence indicator, ACS, Reag. Ph Eur

Synonym(s):

2′,7′-Dichlorofluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Cl2O5
CAS Number:
Molecular Weight:
401.20
MDL number:
UNSPSC Code:
12171501
EC Index Number:
200-968-6
NACRES:
NA.21

product name

2′,7′-Dichlorofluorescein, indicator ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

loss

≤3% loss on drying, 110°C

bulk density

390 kg/m3

storage temp.

2-30°C

InChI

1S/C20H10Cl2O5/c21-13-5-11-17(7-15(13)23)26-18-8-16(24)14(22)6-12(18)20(11)10-4-2-1-3-9(10)19(25)27-20/h1-8,23-24H

InChI key

VFNKZQNIXUFLBC-UHFFFAOYSA-N

Analysis Note

Clarity of solution (according to ACS): passes test
Loss on drying (110 °C): ≤ 3
Suitability as adsorption indicator (according to ACS): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service