Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07174

SAFC

Paraffin

highly liquid, Ph. Eur., BP, JP, NF

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181503
EC Index Number:
232-455-8

Agency

BP
JP
NF
Ph. Eur.

Quality Level

vapor pressure

0.0001 hPa ( 20 °C)

form

liquid

autoignition temp.

>300 °C

kinematic viscosity

16 cSt(40 °C)

bp

300-450 °C/1,013 hPa

transition temp

flash point >180 °C

density

0.85 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

2-30°C

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Paraffin highly liquid is suitable for use in pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

275.0 °F

Flash Point(C)

135 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service