Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.04003

Sigma-Aldrich

Formaldehyde solution about 37%

GR for analysis stabilized with about 10% methanol ACS,Reag. Ph Eur

Synonym(s):

Formaldehyde solution, Formaline solution, Methanal solution, Methylaldehyde solution, Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12352114
NACRES:
NA.21
grade:
ACS reagent
application(s):
diagnostic assay manufacturing
hematology
histology
bp:
93-96 °C/1013 hPa
vapor pressure:
52 mmHg ( 37 °C)

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

300 °C (Formaldehyde)
572 °F

expl. lim.

7-73 % (v/v) Formaldehyde)

concentration

36.5-38.0% (iodometric, acidimetric)

pH

2.8-4.0 (20 °C in H2O)

bp

93-96 °C/1013 hPa

mp

<-15 °C

transition temp

flash point 62 °C (Formaldehyde)

density

1.09 g/cm3 at 20 °C
1.09 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde (HCOH) is one of the most familiar and effective fixatives. The fixation of the histological specimen material of human or animal origin is commonly performed in a 4% or 10% Formaldehyde solution. Chemically, the 4 % Formaldehyde solution equals a 10 % Formalin solution.

Analysis Note

Assay (acidimetric): 36.5 - 38.0 %
Assay (iodometric): 36.5 - 38.0 %
Identity: passes test
Color: ≤ 10 Hazen
Appearance of solution: passes test
Free acid (as HCOOH): ≤ 0.025 %
Density (20 °C): 1.080 - 1.090 g/ml
Chloride (Cl): ≤ 0.0001 %
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.0002 %
Fe (Iron): ≤ 0.0001 %
Methanol (GC): 9.0 - 11.0 %
Sulfated ash: ≤ 0.002 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Preparation of a standard solution for Formaldehyde

Related Content

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service