Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02263

Supelco

Cerium(IV) oxide

1-3 mm for elementary analysis

Synonym(s):

Cerium(IV) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2
CAS Number:
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-150-4
NACRES:
NA.21

Quality Level

form

solid

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

2000 °C

density

7.3 g/cm3 at 20 °C

bulk density

1280 kg/m3

cation traces

Fe: ≤0.02

storage temp.

no temp limit

SMILES string

O=[Ce]=O

InChI

1S/Ce.2O

InChI key

CETPSERCERDGAM-UHFFFAOYSA-N

Application


  • Application of copper (I) selective ligands for PET imaging of reactive oxygen species through metabolic trapping: Bathocuproinedisulfonic acid disodium salt is utilized in the synthesis of copper (I) selective ligands. These ligands are essential for PET imaging to detect reactive oxygen species, highlighting its role in advanced medical imaging and research. This application is pivotal in understanding cellular mechanisms and disease pathologies where oxidative stress is involved (Tada, Mizuno, Shibata, Yasui, Kuge, 2024).

Analysis Note

Sulphate (SO₄): ≤ 0.5 %
Fe (Iron): ≤ 0.02
Loss on ignition (1000 °C): ≤ 1.0 %
Particle size (1-3.15 mm): ≥ 80 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service