Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.00630

Supelco

Nitric acid solution

c(HNO) = 10 mol/l (10 N) for acidification

Synonym(s):

Nitric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

product name

Nitric acid, c(HNO) = 10 mol/l (10 N)

vapor pressure

9.4 hPa ( 20 °C)

Quality Level

product line

Titripur®

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

10 M

technique(s)

titration: suitable

pH

<1 (20 °C in H2O)

bp

120 °C/1013 hPa

mp

-28 °C

density

1.38 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Structural, Electrical, and Optical Properties of Single-Walled Carbon Nanotubes Synthesized through Floating Catalyst Chemical Vapor Deposition.: This study explores the production and analysis of single-walled carbon nanotubes using nitric acid in the synthesis process, highlighting advancements in nanotechnology and material science (Dolafi Rezaee et al., 2024).

  • Separation of gold from irradiated actinide material utilizing a 2-thenoyltrifluoroacetone extraction resin.: This paper discusses the utilization of nitric acid in the separation of gold from actinide materials, demonstrating a significant application in the field of analytical chemistry (Warzecha et al., 2024).

  • Production and separation of (7)Be for use in an ion source.: This article covers the production and separation techniques involving nitric acid, contributing to developments in radiochemistry and isotopic applications (Satija et al., 2024).

  • Optimal production and purification of n.c.a.(143)Pr as a promising palliative agent for the treatment of metastatic bone pain.: While focusing on medical applications, this study uses nitric acid in the synthesis and purification process, highlighting its versatility across different fields (Shirvani-Arani et al., 2024).

Analysis Note

Assay (HNO₃): 48.0 - 50.0 %

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service