Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00525

Millipore

Tryptic Soy Broth - Dehydrated Culture Media

GranuCult® plus, USP, EP, granular, from non-animal source, suitable for microbiology

Synonym(s):

TSB, soy broth, CASO Broth, Casein-peptone Soymeal-peptone Broth, CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106213
NACRES:
NA.74

biological source

non-animal source

Quality Level

Agency

EP
USP

sterility

non-sterile

form

granular

packaging

pkg of 25 kg
pkg of 5 kg

manufacturer/tradename

GranuCult® plus

storage condition

(Tightly closed. Dry)

technique(s)

microbiological culture: suitable

color

beige

pH

7.1-7.5 (25 °C, 30 g/L in H2O, after autoclaving)

solubility

30 g/L

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Cronobacter spp. (spp.)
Escherichia coli (pathogenic)
Salmonella spp. (spp.)
anaerobic bacteria
nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth is a universal culture medium free from inhibitors and indicators for a wide spectrum of applications. The Peptones used in the broth are of non-animal origin. The formulation of the basic medium is prepared according to the recommendations of the current European and the United States Pharmacopoeia (EP, 2.6.12. and USP, 61) but contains peptones of non-animal origin instead of the recommended peptones.

Application

Tryptic Soy Broth is suitable for the isolation and cultivation of fastidious aerobic bacteria, yeasts, and molds. The medium is used for sterility testing of substances, preparations, and products as well as a pre-enrichment broth for the test for specified microorganisms.

Features and Benefits

  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment.
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • A high number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Linkage

Replaces: 1005259027

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service