Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-007

Sigma-Aldrich

Anti-acetyl STAT5B (Lys694) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Signal transducer and activator of transcription 5B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (immunogen homology)

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys694)

Gene Information

human ... GALNT16(57452)

General description

STAT5B (signal transducers and activators of transcription 5b) controls the expression of early response genes and takes part in cytokine signaling. Studies have linked this protein to the inhibition of NFkappaB-mediated signaling. Additional studies have shown an association of STAT5B with regulation of lactation and growth, performance, and control of adipocyte function. Constitutively active STAT5B expression is often observed in a large number of leukemia’s and solid tumors.

Specificity

This antibody recognizes STAT5B acetylated at Lys694.

Immunogen

Epitope: Acetylated Lys694
KLH-conjugated linear peptide corresponding to mouse STAT5B acetylated at Lys694.

Application

Anti-acetyl STAT5B (Lys694) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl STAT5B (Lys694) also known as Signal transducer & activator of transcription 5B & has been validated in DB & WB.

Quality

Evaluated by Dot Blot in modified and non modified peptides of acetylated and non-acetylated STAT5B protein.

Dot Blot Analysis: 1 µg/mL of this antibody detected STAT5B in modified and non modified peptides of acetylated and non-acetylated STAT5B protein.

Target description

90 kDa calculated

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service