Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-2130

Sigma-Aldrich

Anti-Ubiquitin Antibody

from rabbit

Synonym(s):

Polyubiquitin-C, Ubiquitin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBC(7316)

General description

Ubiquitin functions as a marker to indicate the fate of other proteins in the cell. It can be attached as a single molecule or as a polyubiquitin chain. Although ubiquitinated proteins are oftentimes tagged for degradation by the 20S proteosome, ubiquitination can also indicate alternative fates depending on the lysine residue that is involved in linking the ubiquitin chain to the target protein.Ubiquitin chains linked by Lys33 to kinase substrates such as AMP-activated protein kinases, (AMPKα1, BRSK1, MARK3, MARK4, and NUAK1), may activate or deactivate these proteins, supporting the role for uniquitin in regulating signaling.

Specificity

Broad species reactivity expected based on sequence similarity.
This antibody recognizes Ubiquitin at the N-terminus

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human Ubiquitin.

Application

Research Category
Signaling
Research Sub Category
General Post-translation Modification
This Anti-Ubiquitin antibody is validated for use in IP & WB for the detection of the Ubiquitin protein.
Western Blot Analysis: 0.5 µg/mL from a representative lot detected Ubiquitin on 10 µg of HeLa cell lysate.

Quality

Evaluated by Immunoprecipitation in serum starved HeLa cell lysate.

Immunoprecipitation Analysis: 10 µg of this antibody detected Ubiquitin on 5 µL of serum starved HeLa cell lysate.

Target description

~8 kDa observed. The calculated molecular weight of this protein is 8 kDa, but has been observed at ~24 kDa due to tetramers. (Kvint, K., et al. (2008). Mol Cell.30(4):498-506.)

Physical form

Affinity purified
Format: Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Serum starved HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristian Kvint et al.
Molecular cell, 30(4), 498-506 (2008-05-24)
The final outcome of protein polyubiquitylation is often proteasome-mediated proteolysis, meaning that "proofreading" of ubiquitylation by ubiquitin proteases (UBPs) is crucial. Transcriptional arrest can trigger ubiquitin-mediated proteolysis of RNA polymerase II (RNAPII) so a UBP reversing RNAPII ubiquitylation might be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service