Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-468

Sigma-Aldrich

Anti-FcεRI α subunit Antibody

Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

rat

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FCER1A(2205)

Specificity

FcεRIα

Immunogen

Whole rat basophilic leukemia cells, positive for the rat high-affinity receptor recognizing immunoglobulin E (FcεRI).

Application

Anti-FcεRI α subunit Antibody is an antibody against FcεRI α subunit for use in FC & IP.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
Western blot is not recommended

Quality

routinely evaluated by immunoprecipitation of rat FcεRIα from biotinylated rat basophilic leukemia cell lysate

Target description

45-55kDa

Physical form

0.1M Tris-glycine, 0.15M NaCl, 0.05% sodium azide prior to the addition of glycerol to 30%
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycosylation of human truncated Fc epsilon RI alpha chain is necessary for efficient folding in the endoplasmic reticulum.
Letourneur, O, et al.
The Journal of Biological Chemistry, 270, 8249-8256 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service