Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

03-34-0035

Sigma-Aldrich

H-Gly-Arg-Gly-Asp-Ser-Pro-OH

Shown to inhibit fibronectin binding to platelet-binding sites.

Synonym(s):

H-Gly-Arg-Gly-Asp-Ser-Pro-OH, GRGDSP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H37N9O10
Molecular Weight:
587.58
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

5% acetic acid: 1 mg/mL
water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Shown to inhibit fibronectin binding to platelet-binding sites.

Biochem/physiol Actions

Cell permeable: no
Primary Target
fibronectin binding
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Pierschbacher, M.D., and Ruoslahti, E. 1984. Nature309, 30.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service